C12H316CE4A5 Chemcia Scientific, LLC

2-Methyl-4-oxo-3,5,7,8-tetrahydro-4H-pyrido[4,3-d]pyrimidine-6-carboxylic acid tert-butyl ester

  • MDL number MFCD22395983
  • Cas Number : 1073440-84-2
  • Molecular Weight : 265.31468

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Beige Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1099

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1099
Availability details for
Quantity
  • Availability
  • Add to Cart