C12H316CE4AE Chemcia Scientific, LLC

2-Bromo-1H-indole-3-carbaldehyde

  • MDL number MFCD18449874
  • Cas Number : 119910-45-1
  • Molecular Weight : 224.05827

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1091

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1091
Availability details for
Quantity
  • Availability
  • Add to Cart