C12H316CE4B1 Chemcia Scientific, LLC

2-Amino-4-oxo-3,5,7,8-tetrahydro-4H-pyrido[4,3-d]pyrimidine-6-carboxylic acid tert-butyl ester

  • MDL number MFCD09999172
  • Cas Number : 1000386-01-5
  • Molecular Weight : 266.30226

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Pink Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1088

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1088
Availability details for
Quantity
  • Availability
  • Add to Cart