C12H316CE4B6 Chemcia Scientific, LLC

2-(2-Nitro-benzylidene)-malonic acid diethyl ester

  • MDL number MFCD02176752
  • Cas Number : 17422-56-9
  • Molecular Weight : 293.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name diethyl 2-(2-nitrobenzylidene)malonate
Inchi Code 1S/C14H15NO6/c1-3-20-13(16)11(14(17)21-4-2)9-10-7-5-6-8-12(10)15(18)19/h5-9H,3-4H2,1-2H3
Inchi Key BLHLIGAACQEKRY-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. 0~5
Physical Form Yellow Crystalline Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1076

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1076
Availability details for
Quantity
  • Availability
  • Add to Cart