C12H316CE4C4 Chemcia Scientific, LLC

N-(2-Bromo-5-fluoro-phenyl)-formamide

  • MDL number MFCD22565617
  • Cas Number : 1395493-19-2
  • Molecular Weight : 218.0264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1793

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1793
Availability details for
Quantity
  • Availability
  • Add to Cart