C12H316CE4CD Chemcia Scientific, LLC

6-Methylimidazo[1,2-a]pyridine

  • MDL number MFCD05663806
  • Cas Number : 874-38-4
  • Molecular Weight : 132.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methylimidazo[1,2-a]pyridine
Inchi Code 1S/C8H8N2/c1-7-2-3-8-9-4-5-10(8)6-7/h2-6H,1H3
Inchi Key VKVFDPQDGZSLJK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1152

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1152
Availability details for
Quantity
  • Availability
  • Add to Cart