C12H316CE4CF Chemcia Scientific, LLC

6-Benzyloxy-4-oxo-1,4-dihydro-quinoline-2-carboxylic acid methyl ester

  • MDL number MFCD22565618
  • Cas Number : 929028-73-9
  • Molecular Weight : 309.32455

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1179

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1179
Availability details for
Quantity
  • Availability
  • Add to Cart