C12H316CE4D1 Chemcia Scientific, LLC

2-Amino-3-methyl-3H-pyrrolo[3,2-d]pyrimidin-4(5H)-one

  • MDL number MFCD09540492
  • Cas Number : 151587-61-0
  • Molecular Weight : 164.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-3-methyl-3,5-dihydro-4H-pyrrolo[3,2-d]pyrimidin-4-one
Inchi Code 1S/C7H8N4O/c1-11-6(12)5-4(2-3-9-5)10-7(11)8/h2-3,9H,1H3,(H2,8,10)
Inchi Key WUCZWUCLYHUJPL-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1366

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1366
Availability details for
Quantity
  • Availability
  • Add to Cart