C12H316CE4D6 Chemcia Scientific, LLC

6-Nitro-8-bromo-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester

  • MDL number MFCD19441958
  • Cas Number : 1313712-72-9
  • Molecular Weight : 314.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 8-bromo-6-nitroimidazo[1,2-a]pyridine-2-carboxylate
Inchi Code 1S/C10H8BrN3O4/c1-2-18-10(15)8-5-13-4-6(14(16)17)3-7(11)9(13)12-8/h3-5H,2H2,1H3
Inchi Key KUKFEJRSKIUIIC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1073

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1073
Availability details for
Quantity
  • Availability
  • Add to Cart