C12H316CE4DD Chemcia Scientific, LLC

2-Chloro-quinoline-3-carboxylic acid ethyl ester

  • MDL number MFCD06204685
  • Cas Number : 16498-86-5
  • Molecular Weight : 235.672

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1143

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1143
Availability details for
Quantity
  • Availability
  • Add to Cart