C12H316CE4E0 Chemcia Scientific, LLC

3-Nitro-1-methyl-2(1H)-pyridinone

  • MDL number MFCD01646143
  • Cas Number : 32896-91-6
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-methyl-3-nitro-2(1H)-pyridinone
Inchi Code 1S/C6H6N2O3/c1-7-4-2-3-5(6(7)9)8(10)11/h2-4H,1H3
Inchi Key BDQAZNBCEXTXIW-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1125

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1125
Availability details for
Quantity
  • Availability
  • Add to Cart