C12H316CE4E1 Chemcia Scientific, LLC

Pyridin-4-yl-acetic acid tert-butyl ester

  • MDL number MFCD22565620
  • Cas Number : 79757-20-3
  • Molecular Weight : 193.2477

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1618

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1618
Availability details for
Quantity
  • Availability
  • Add to Cart