C12H316CE4E2 Chemcia Scientific, LLC

8-Bromo-6-nitroimidazo[1,2-a]pyridine

  • MDL number MFCD16619380
  • Cas Number : 957187-28-9
  • Molecular Weight : 242.03283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1126

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1126
Availability details for
Quantity
  • Availability
  • Add to Cart