C12H316CE4E3 Chemcia Scientific, LLC

N-(1H-Pyrazol-3-yl)acetimidamide

  • MDL number MFCD22565621
  • Cas Number : 51247-95-1
  • Molecular Weight : 124.14631

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1144

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1144
Availability details for
Quantity
  • Availability
  • Add to Cart