C12H316CE4E8 Chemcia Scientific, LLC

4-Chloro-5-(chlorosulfonyl)-2-methoxybenzoic acid

  • MDL number MFCD22565622
  • Cas Number : 57479-73-9
  • Molecular Weight : 285.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-5-(chlorosulfonyl)-2-methoxybenzoic acid
Inchi Code 1S/C8H6Cl2O5S/c1-15-6-3-5(9)7(16(10,13)14)2-4(6)8(11)12/h2-3H,1H3,(H,11,12)
Inchi Key KGGQSUTYQMJRNQ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1148

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1148
Availability details for
Quantity
  • Availability
  • Add to Cart