C12H316CE4EF Chemcia Scientific, LLC

3-Bromo-6-chloro-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester hydrobromide

  • MDL number MFCD22570219
  • Cas Number : 1951441-94-3
  • Molecular Weight : 384.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate hydrobromide
Inchi Code 1S/C10H8BrClN2O2.BrH/c1-2-16-10(15)8-9(11)14-5-6(12)3-4-7(14)13-8;/h3-5H,2H2,1H3;1H
Inchi Key BYSBIKPVWZRGEN-UHFFFAOYSA-N

Description

Purity 95%
Salt Data HBr
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1370

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1370
Availability details for
Quantity
  • Availability
  • Add to Cart