C12H316CE4F2 Chemcia Scientific, LLC

6-Chloro-4-iodopyridin-3-ylamine hydrochloride

  • MDL number MFCD22570214
  • Molecular Weight : 290.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-4-iodopyridin-3-amine hydrochloride
Inchi Code 1S/C5H4ClIN2.ClH/c6-5-1-3(7)4(8)2-9-5;/h1-2H,8H2;1H
Inchi Key QDDAWXSBSWOTPQ-UHFFFAOYSA-N

Description

Purity 97%
Salt Data HCl
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1231

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1231
Availability details for
Quantity
  • Availability
  • Add to Cart