C12H316CE4F6 Chemcia Scientific, LLC

2-Amino-5-iodo-6-methylpyrimidin-4(3H)-one

  • MDL number MFCD21649213
  • Cas Number : 22294-57-1
  • Molecular Weight : 251.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-5-iodo-6-methyl-4-pyrimidinol
Inchi Code 1S/C5H6IN3O/c1-2-3(6)4(10)9-5(7)8-2/h1H3,(H3,7,8,9,10)
Inchi Key LIZYQAUQOBJSBC-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1234

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1234
Availability details for
Quantity
  • Availability
  • Add to Cart