C12H316CE4FA Chemcia Scientific, LLC

(5-Chloro-pyridin-2-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD11504986
  • Cas Number : 354503-86-9
  • Molecular Weight : 228.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 5-chloro-2-pyridinylcarbamate
Inchi Code 1S/C10H13ClN2O2/c1-10(2,3)15-9(14)13-8-5-4-7(11)6-12-8/h4-6H,1-3H3,(H,12,13,14)
Inchi Key CEZBERPWXUYBQB-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1238

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1238
Availability details for
Quantity
  • Availability
  • Add to Cart