C12H316CE4FD Chemcia Scientific, LLC

5-Pyridin-4-yl-2,4-dihydro-[1,2,4]triazol-3-one

  • MDL number MFCD19685619
  • Cas Number : 939-08-2
  • Molecular Weight : 162.15207

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Khaki Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1241

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1241
Availability details for
Quantity
  • Availability
  • Add to Cart