C12H316CE500 Chemcia Scientific, LLC

2-tert-Butoxycarbonylamino-4,5,6,7-tetrahydro-benzothiazole-4-carboxylic acid ethyl ester

  • MDL number MFCD22565627
  • Cas Number : 1369897-34-6
  • Molecular Weight : 326.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-((tert-butoxycarbonyl)amino)-4,5,6,7-tetrahydrobenzo[d]thiazole-4-carboxylate
Inchi Code 1S/C15H22N2O4S/c1-5-20-12(18)9-7-6-8-10-11(9)16-13(22-10)17-14(19)21-15(2,3)4/h9H,5-8H2,1-4H3,(H,16,17,19)
Inchi Key SWYBGWIBKLDLCP-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1244

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1244
Availability details for
Quantity
  • Availability
  • Add to Cart