C12H316CE506 Chemcia Scientific, LLC

2-(3-Methyl-2-nitrophenyl)acetonitrile

  • MDL number MFCD08276005
  • Cas Number : 91192-25-5
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-methyl-2-nitrophenyl)acetonitrile
Inchi Code 1S/C9H8N2O2/c1-7-3-2-4-8(5-6-10)9(7)11(12)13/h2-4H,5H2,1H3
Inchi Key QRUQKLCGZUEVHE-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1249

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1249
Availability details for
Quantity
  • Availability
  • Add to Cart