C12H316CE509 Chemcia Scientific, LLC

8-Bromo-6-chloro-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester

  • MDL number MFCD09607953
  • Cas Number : 951884-22-3
  • Molecular Weight : 303.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 8-bromo-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Inchi Code 1S/C10H8BrClN2O2/c1-2-16-10(15)8-5-14-4-6(12)3-7(11)9(14)13-8/h3-5H,2H2,1H3
Inchi Key FOHQDZNEJCDSSN-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1251

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1251
Availability details for
Quantity
  • Availability
  • Add to Cart