C12H316CE514 Chemcia Scientific, LLC

6-Fluoro-4-oxo-1,4-dihydro-quinoline-2-carboxylic acid methyl ester

  • MDL number MFCD11998328
  • Cas Number : 19271-19-3
  • Molecular Weight : 221.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 6-fluoro-4-oxo-1,4-dihydro-2-quinolinecarboxylate
Inchi Code 1S/C11H8FNO3/c1-16-11(15)9-5-10(14)7-4-6(12)2-3-8(7)13-9/h2-5H,1H3,(H,13,14)
Inchi Key BESDSRIEDPEFRN-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1259

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1259
Availability details for
Quantity
  • Availability
  • Add to Cart