C12H316CE517 Chemcia Scientific, LLC

2-tert-Butoxycarbonylamino-4,5,6,7-tetrahydro-benzothiazole-4-carboxylic acid

  • MDL number MFCD22565633
  • Cas Number : 1190391-84-4
  • Molecular Weight : 298.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-((tert-butoxycarbonyl)amino)-4,5,6,7-tetrahydrobenzo[d]thiazole-4-carboxylic acid
Inchi Code 1S/C13H18N2O4S/c1-13(2,3)19-12(18)15-11-14-9-7(10(16)17)5-4-6-8(9)20-11/h7H,4-6H2,1-3H3,(H,16,17)(H,14,15,18)
Inchi Key QQAPPTIFMLODJN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White-like Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1365

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1365
Availability details for
Quantity
  • Availability
  • Add to Cart