C12H316CE519 Chemcia Scientific, LLC

3-Hydroxy-7,8-dihydro-5H-pyrido[4,3-c]pyridazine-6-carboxylic acid tert-butyl ester

  • MDL number MFCD07779188
  • Cas Number : 890091-87-9
  • Molecular Weight : 251.28759

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1167

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1167
Availability details for
Quantity
  • Availability
  • Add to Cart