C12H316CE520 Chemcia Scientific, LLC

6-Chloro-4-oxo-1,4-dihydro-quinoline-2-carboxylic acid methyl ester

  • MDL number MFCD07801026
  • Cas Number : 7545-52-0
  • Molecular Weight : 237.64431

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1268

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1268
Availability details for
Quantity
  • Availability
  • Add to Cart