C12H316CE525 Chemcia Scientific, LLC

6,7-Dihydro-2H-pyrrolo[3,4-c]pyridazin-3(5H)-one hydrochloride

  • MDL number MFCD22570211
  • Cas Number : 1415550-33-2
  • Molecular Weight : 173.60316

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Salt Data HCl
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1161

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1161
Availability details for
Quantity
  • Availability
  • Add to Cart