C12H316CE526 Chemcia Scientific, LLC

6,7-Dihydro-5H-pyrrolo[3,4-c]pyridazin-3-ol hydrochloride

  • MDL number MFCD22570212
  • Cas Number : 1415550-33-2
  • Molecular Weight : 173.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6,7-dihydro-5H-pyrrolo[3,4-c]pyridazin-3-ol hydrochloride
Inchi Code 1S/C6H7N3O.ClH/c10-6-1-4-2-7-3-5(4)8-9-6;/h1,7H,2-3H2,(H,9,10);1H
Inchi Key CKVFKGRDRZJGNR-UHFFFAOYSA-N

Description

Purity 95%
Salt Data HCl
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1162

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1162
Availability details for
Quantity
  • Availability
  • Add to Cart