C12H316CE527 Chemcia Scientific, LLC

5-Fluoro-3-methoxy-2-nitrobenzoic acid

  • MDL number MFCD22565642
  • Cas Number : 1007113-04-3
  • Molecular Weight : 215.13912

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1163

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1163
Availability details for
Quantity
  • Availability
  • Add to Cart