C12H316CE52E Chemcia Scientific, LLC

5-(Dimethylamino)-2-(2,2,2-trifluoroacetyl)penta-2,4-dienenitrile

  • MDL number MFCD22565645
  • Cas Number : 1022901-66-1
  • Molecular Weight : 218.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2E,4E)-5-(dimethylamino)-2-(2,2,2-trifluoroacetyl)penta-2,4-dienenitrile
Inchi Code 1S/C9H9F3N2O/c1-14(2)5-3-4-7(6-13)8(15)9(10,11)12/h3-5H,1-2H3/b5-3+,7-4+
Inchi Key QEBYTZVFLQLFRY-HJIKTHEYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Red Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1276

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1276
Availability details for
Quantity
  • Availability
  • Add to Cart