C12H316CE535 Chemcia Scientific, LLC

3,5-Difluoro-2-nitrobenzonitrile

  • MDL number MFCD13185568
  • Cas Number : 868596-13-8
  • Molecular Weight : 184.10299

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1281

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1281
Availability details for
Quantity
  • Availability
  • Add to Cart