C12H316CE53A Chemcia Scientific, LLC

2-(2-Aminobenzo[d]thiazol-6-yl)acetonitrile

  • MDL number MFCD11844845
  • Cas Number : 352530-05-3
  • Molecular Weight : 189.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-amino-1,3-benzothiazol-6-yl)acetonitrile
Inchi Code 1S/C9H7N3S/c10-4-3-6-1-2-7-8(5-6)13-9(11)12-7/h1-2,5H,3H2,(H2,11,12)
Inchi Key KAUYEYJEFMZZGK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1286

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1286
Availability details for
Quantity
  • Availability
  • Add to Cart