C12H316CE541 Chemcia Scientific, LLC

5,6,7,8-Tetrahydrocinnolin-3-ol

  • MDL number MFCD20502835
  • Cas Number : 5468-36-0
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5,6,7,8-tetrahydro-3-cinnolinol
Inchi Code 1S/C8H10N2O/c11-8-5-6-3-1-2-4-7(6)9-10-8/h5H,1-4H2,(H,10,11)
Inchi Key YZVBTQDJTRCXMW-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1293

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1293
Availability details for
Quantity
  • Availability
  • Add to Cart