C12H316CE547 Chemcia Scientific, LLC

(6-Methylimidazo[1,2-a]pyridin-2-yl)methanol

  • MDL number MFCD08442125
  • Cas Number : 926223-25-8
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-methylimidazo[1,2-a]pyridin-2-yl)methanol
Inchi Code 1S/C9H10N2O/c1-7-2-3-9-10-8(6-12)5-11(9)4-7/h2-5,12H,6H2,1H3
Inchi Key AYLQQEGSDYADDW-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1299

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1299
Availability details for
Quantity
  • Availability
  • Add to Cart