C12H316CE549 Chemcia Scientific, LLC

6-Methoxy-2-methyl-1H-indole

  • MDL number MFCD09038152
  • Cas Number : 1968-13-4
  • Molecular Weight : 161.20527

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1301

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1301
Availability details for
Quantity
  • Availability
  • Add to Cart