C12H316CE54E Chemcia Scientific, LLC

2-Bromo-4-methoxy-6-nitroaniline

  • MDL number MFCD18088521
  • Cas Number : 10172-35-7
  • Molecular Weight : 247.04944

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Red Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1305

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1305
Availability details for
Quantity
  • Availability
  • Add to Cart