C12H316CE54F Chemcia Scientific, LLC

N-(4-Cyanomethyl-2-nitro-phenyl)-acetamide

  • MDL number MFCD00189257
  • Cas Number : 123270-23-5
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-[4-(cyanomethyl)-2-nitrophenyl]acetamide
Inchi Code 1S/C10H9N3O3/c1-7(14)12-9-3-2-8(4-5-11)6-10(9)13(15)16/h2-3,6H,4H2,1H3,(H,12,14)
Inchi Key XHJXYBIIUUFVLB-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1306

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1306
Availability details for
Quantity
  • Availability
  • Add to Cart