C12H316CE555 Chemcia Scientific, LLC

2-Cyclopropyl-5,7-dihydro-pyrrolo[3,4-d]pyrimidine-6-carboxylic acid tert-butyl ester

  • MDL number In Stock
  • Cas Number : 1289267-20-4
  • Molecular Weight : 261.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-cyclopropyl-5,7-dihydro-6H-pyrrolo[3,4-d]pyrimidine-6-carboxylate
Inchi Code 1S/C14H19N3O2/c1-14(2,3)19-13(18)17-7-10-6-15-12(9-4-5-9)16-11(10)8-17/h6,9H,4-5,7-8H2,1-3H3
Inchi Key KLKHYMNNVCROMT-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1189

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1189
Availability details for
Quantity
  • Availability
  • Add to Cart