C12H316CE556 Chemcia Scientific, LLC

2-Cyclopropyl-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidine hydrochloride

  • MDL number MFCD22570225
  • Molecular Weight : 211.69618

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Salt Data HCl
Storage temp. 0~5
Physical Form Pale Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1183

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1183
Availability details for
Quantity
  • Availability
  • Add to Cart