C12H316CE55D Chemcia Scientific, LLC

6-Acetylamino-4-oxo-1,4-dihydro-quinoline-2-carboxylic acid methyl ester

  • MDL number MFCD11855916
  • Cas Number : 929028-74-0
  • Molecular Weight : 260.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 6-(acetylamino)-4-oxo-1,4-dihydro-2-quinolinecarboxylate
Inchi Code 1S/C13H12N2O4/c1-7(16)14-8-3-4-10-9(5-8)12(17)6-11(15-10)13(18)19-2/h3-6H,1-2H3,(H,14,16)(H,15,17)
Inchi Key JNZLYYAFMTXECK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1316

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1316
Availability details for
Quantity
  • Availability
  • Add to Cart