C12H316CE560 Chemcia Scientific, LLC

6-(6-Amino-hexyldisulfanyl)-hexylamine dihydrochloride

  • MDL number MFCD22570213
  • Cas Number : 118077-46-6
  • Molecular Weight : 337.4203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0~5
Physical Form Pale Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1184

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1184
Availability details for
Quantity
  • Availability
  • Add to Cart