C12H316CE562 Chemcia Scientific, LLC

6-Amino-4-oxo-1,4-dihydro-quinoline-2-carboxylic acid methyl ester

  • MDL number MFCD01934583
  • Cas Number : 52979-42-7
  • Molecular Weight : 218.21395

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1190

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1190
Availability details for
Quantity
  • Availability
  • Add to Cart