C12H316CE56A Chemcia Scientific, LLC

5-Bromo-pyridine-2,3-dicarboxylic acid dimethyl ester

  • MDL number MFCD09953248
  • Cas Number : 521980-82-5
  • Molecular Weight : 274.07241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1323

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1323
Availability details for
Quantity
  • Availability
  • Add to Cart