C12H316CE56F Chemcia Scientific, LLC

2-Chloro-6-methoxybenzonitrile

  • MDL number MFCD00019743
  • Molecular Weight : 167.59612

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1328

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1328
Availability details for
Quantity
  • Availability
  • Add to Cart