C12H316CE573 Chemcia Scientific, LLC

5-Bromo-2,3-bis-chloromethyl-pyridine

  • MDL number MFCD18262035
  • Cas Number : 155187-02-3
  • Molecular Weight : 254.94257

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1374

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1374
Availability details for
Quantity
  • Availability
  • Add to Cart