C12H316CE577 Chemcia Scientific, LLC

2-Methyl-5,6,7,8-tetrahydro-pyrido[4,3-d]pyrimidine hydrochloride

  • MDL number MFCD11043222
  • Cas Number : 210538-73-1
  • Molecular Weight : 185.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidine hydrochloride
Inchi Code 1S/C8H11N3.ClH/c1-6-10-5-7-4-9-3-2-8(7)11-6;/h5,9H,2-4H2,1H3;1H
Inchi Key ORJSIFYUPXLPLK-UHFFFAOYSA-N

Description

Purity 97%
Salt Data HCl
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1193

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1193
Availability details for
Quantity
  • Availability
  • Add to Cart