C12H316CE578 Chemcia Scientific, LLC

(3-Bromo-4-methyl-phenyl)-carbamic acid tert-butyl ester

  • MDL number MFCD12913704
  • Cas Number : 515813-02-2
  • Molecular Weight : 286.17082

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1335

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1335
Availability details for
Quantity
  • Availability
  • Add to Cart