C12H316CE579 Chemcia Scientific, LLC

(3-Bromo-4-methyl-phenyl)-methyl-carbamic acid tert-butyl ester

  • MDL number MFCD12913705
  • Cas Number : 877064-95-4
  • Molecular Weight : 300.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-bromo-4-methylphenyl(methyl)carbamate
Inchi Code 1S/C13H18BrNO2/c1-9-6-7-10(8-11(9)14)15(5)12(16)17-13(2,3)4/h6-8H,1-5H3
Inchi Key RQGCJQIAGDDTKJ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Pale Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1336

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1336
Availability details for
Quantity
  • Availability
  • Add to Cart