C12H316CE57A Chemcia Scientific, LLC

(3-Bromo-4-methyl-phenyl)-methyl-amine

  • MDL number MFCD09745409
  • Cas Number : 101251-11-0
  • Molecular Weight : 200.0796

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Pale Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1337

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1337
Availability details for
Quantity
  • Availability
  • Add to Cart